Anti-GPR63 Rabbit Polyclonal Antibody
Catalog # 77704-677
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GPR63
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BZJ6
- Antigen Synonyms:Probable G-protein coupled receptor 63|PSP24-beta|PSP24-2|PSP24B|G protein-coupled receptor 63|PSP24(beta) |Brain expressed G-protein-coupled receptor PSP24 beta|GPR63_HUMAN|Gpr63|G Protein-Coupled Receptor GPR63
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human GPR63
- Purification:Antigen affinity purified
- Cat. No.:77704-677
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: GPR63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse