Anti-OR4F17/OR4F4/OR4F5 Rabbit Polyclonal Antibody
Catalog # 77704-667
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OR4F17/OR4F4/OR4F5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NGA8/Q96R69/Q8NH21
- Antigen Synonyms:HS14a-1-A|Olfactory receptor 4F5|Olfactory receptor 4F11|Olfactory receptor 4F18|OR4F11P|OR4F18|Olfactory receptor 4F4|OR4F19|OR4F5|Olfactory receptor OR19-3|OR4F4|OR4F17|Olfactory receptor 4F19|Olfactory receptor 4F17
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Olfactory receptor 4F4/4F5/4F17
- Purification:Antigen affinity purified
- Cat. No.:77704-667
- Packaging:Vial
Specifications
About this item
Type:
Antigen: OR4F17/OR4F4/OR4F5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human