Anti-NCF1 Rabbit Polyclonal Antibody
Catalog # 77704-275
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:NCF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P14598
- Antigen Synonyms:p47-phox|Nox organizing protein 2|NCF1_HUMAN|NCF 47K|p47 phox|Ncf1|SH3 and PX domain containing protein 1A|SH3 and PX domain-containing protein 1A|47 kDa neutrophil oxidase factor|NADPH oxidase organizer 2|neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1)|SH3PXD1A|47 kDa autosomal chronic granulomatous disease protein|Nox-organizing protein 2|Neutrophil cytosol factor 1|NOXO2|NCF-1|Neutrophil cytosolic factor 1|Neutrophil NADPH oxidase factor 1|NCF-47K|Nox organizer 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human p47-phox around the non-phosphorylation site of S328
- Purification:Antigen affinity purified
- Cat. No.:77704-275
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: NCF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat