Anti-RFWD2 Rabbit Polyclonal Antibody
Catalog # 77704-211
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RFWD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NHY2
- Antigen Synonyms:Constitutive photomorphogenesis protein 1 homolog|ring finger and WD repeat domain 2|E3 ubiquitin-protein ligase RFWD2|COP|RING finger and WD repeat domain protein 2|RFWD2_HUMAN|Rfwd2|RING finger protein 200|hCOP1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human COP1 around the non-phosphorylation site of S387
- Purification:Antigen affinity purified
- Cat. No.:77704-211
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RFWD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse