Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-RPS6KB2 (phospho-S423) Rabbit Polyclonal Antibody
Anti-RPS6KB2 (phospho-S423) Rabbit Polyclonal Antibody
Catalog # 77704-146
Supplier:  AFG Bioscience
New Product
undefined
Anti-RPS6KB2 (phospho-S423) Rabbit Polyclonal Antibody
Catalog # 77704-146
Supplier:  AFG Bioscience
Supplier Number:  A40305-100UG
New Product

Specifications

  • Antigen Symbol:
    RPS6KB2 (phospho-S423)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    Q9UBS0
  • Antigen Synonyms:
    p70 S6 kinase beta|p70 ribosomal S6 kinase beta|Serine/threonine-protein kinase 14B|S6K-beta|70 kDa ribosomal protein S6 kinase 2|S6K beta 2|S6K2|P70S6K2|p70-S6K 2|p70 S6KB|SRK|Rps6kb2|Ribosomal protein S6 kinase beta 2|Ribosomal protein S6 kinase B2|p70 S6K-beta|p70(S6K) beta|p70S6Kb|EC 2.7.11.1|S6 kinase related kinase|p70 S6Kbeta|Serine/threonine protein kinase 14 beta |S6K beta|S6K-beta-2|Ribosomal protein S6 kinase 70kDa|Ribosomal protein S6 kinase beta-2|STK14B|p70-beta|polypeptide 2|KS6B2_HUMAN|S6 kinase-related kinase|p70 beta
  • Storage Buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage Temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from Human p70 S6 kinase β around the phosphorylation site of S423
  • Purification:
    Antigen affinity purified
  • Cat. No.:
    77704-146
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: RPS6KB2 (phospho-S423)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse