Anti-RB1 (phospho-S788) Rabbit Polyclonal Antibody
77704-049
New Product
- Antigen Symbol:RB1 (phospho-S788)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P06400
- Antigen Synonyms:Osteosarcoma|RB1|RB_HUMAN|pRb|Retinoblastoma 1|PP105|OSRC|Protein phosphatase 1 regulatory subunit 130|Prepro retinoblastoma associated protein|RB1 gene|Retinoblastoma suspectibility protein|P105RB|GOS563 exon 17 substitution mutation causes premature stop|Rb|Exon 17 tumor GOS561 substitution mutation causes premature stop|pp110|PPP1R130|RB transcriptional corepressor 1|Retinoblastoma-associated protein|p105-Rb
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Rb around the phosphorylation site of S788
- Purification:Antigen affinity purified
- Cat. No.:77704-049
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: RB1 (phospho-S788)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat