Anti-SLC2A4 Rabbit Polyclonal Antibody
Catalog # 77701-337
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SLC2A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P14672
- Antigen Synonyms:Glucose transporter GLUT 4|solute carrier family 2 (facilitated glucose transporter) member 4|Glucose transporter type 4 insulin responsive|SLC 2A4|Solute carrier family 2, facilitated glucose transporter member 4|GLUT-4|SLC2A4|Insulin responsive glucose transporter type 4|Solute carrier family 2 member 4|GLUT 4|GLUT4|Glucose transporter type 4|kug|insulin-responsive|GTR4_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human Glut4
- Purification:Antigen affinity purified
- Cat. No.:77701-337
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: SLC2A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat