Anti-TNFRSF10C Rabbit Polyclonal Antibody
77701-301
New Product
- Antigen Symbol:TNFRSF10C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O14798
- Antigen Synonyms:Antagonist decoy receptor for TRAIL/Apo-2L|TNF-related apoptosis-inducing ligand receptor 3|DCR1|TRID|Decoy receptor 1|TRAIL-R3|Lymphocyte inhibitor of TRAIL|TRAILR3|Decoy TRAIL receptor without death domain|DcR1|CD antigen CD263|LIT|UNQ321/PRO366|TRAIL receptor without an intracellular domain|TNFRSF10C|TRAIL receptor 3|Tumor necrosis factor receptor superfamily member 10C
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human DcR1
- Purification:Antigen affinity purified
- Cat. No.:77701-301
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: TNFRSF10C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human