Anti-FAIM2 Rabbit Polyclonal Antibody
Catalog # 77700-909
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FAIM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BWQ8
- Antigen Synonyms:KIAA0950|NGP35|Transmembrane BAX inhibitor motif containing 2|FAIM 2|Faim2|TMBIM 2|NGP 35|Protein lifeguard|FAIM2_HUMAN|NMP35|Fas apoptotic inhibitory molecule 2|TMBIM2|Transmembrane BAX inhibitor motif-containing protein 2|LFG|Transmembrane BAX inhibitor motif containing protein 2|NMP 35|Lifeguard|Neural membrane protein 35
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human FAIM2
- Purification:Antigen affinity purified
- Cat. No.:77700-909
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: FAIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human