Anti-NCK2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: NCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77708-732
- 77708-731
- Antigen Symbol:
- NCK2
- NCK2
- Antigen Synonyms:
- Cytoplasmic protein NCK2|Non-catalytic region of tyrosine kinase adaptor protein 2|NCK adaptor protein 2|NCK2|NCK2_HUMAN|Growth factor receptor bound protein 4|Nck-2|SH2/SH3 adaptor protein NCK-beta|Noncatalytic region of tyrosine kinase beta|GRB4|NCKbeta
- Cytoplasmic protein NCK2|Non-catalytic region of tyrosine kinase adaptor protein 2|NCK adaptor protein 2|NCK2|NCK2_HUMAN|Growth factor receptor bound protein 4|Nck-2|SH2/SH3 adaptor protein NCK-beta|Noncatalytic region of tyrosine kinase beta|GRB4|NCKbeta
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43639
- O43639
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human NCK2
- Synthetic peptide of Human NCK2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.