Anti-AATK Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: AATK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77708-242
- 77708-243
- Antigen Symbol:
- AATK
- AATK
- Antigen Synonyms:
- Brain apoptosis associated tyrosine kinase|AATYK|Serine/threonine protein kinase LMTK1|Apoptosis associated tyrosine kinase|AATK|CDK5-binding protein|Serine/threonine-protein kinase LMTK1|Lemur tyrosine kinase 1|AATYK1|p35BP|LMTK1_HUMAN|p35 binding protein|Brain apoptosis-associated tyrosine kinase|p35-binding protein|LMR1|LMTK1|CDK5 binding protein|Apoptosis-associated tyrosine kinase|KIAA0641
- Brain apoptosis associated tyrosine kinase|AATYK|Serine/threonine protein kinase LMTK1|Apoptosis associated tyrosine kinase|AATK|CDK5-binding protein|Serine/threonine-protein kinase LMTK1|Lemur tyrosine kinase 1|AATYK1|p35BP|LMTK1_HUMAN|p35 binding protein|Brain apoptosis-associated tyrosine kinase|p35-binding protein|LMR1|LMTK1|CDK5 binding protein|Apoptosis-associated tyrosine kinase|KIAA0641
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZMQ8
- Q6ZMQ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human AATK
- Synthetic peptide of Human AATK
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.