Anti-FOLH1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: FOLH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77708-083
- 77708-084
- Antigen Symbol:
- FOLH1
- FOLH1
- Antigen Synonyms:
- mGCP|Folylpoly gamma glutamate carboxypeptidase|Prostate-specific membrane antigen|Folylpoly-gamma-glutamate carboxypeptidase|Pteroylpoly-gamma-glutamate carboxypeptidase|Membrane glutamate carboxypeptidase|GCP2|Cell growth-inhibiting gene 27 protein|FOLH|GCP II|Glutamate carboxypeptidase II|PSM|Folate hydrolase prostate specific membrane antigen 1|NAALAdase|NAALAD 1|GCPII|NAALAD1|N acetylated alpha linked acidic dipeptidase 1|Glutamate carboxylase II|Folate hydrolase (prostate-specific membrane antigen) 1|Prostate specific membrane antigen variant F|Folate hydrolase|PSMA|Cell growth inhibiting protein 27|FOLH 1|Folate hydrolase 1|N-acetylated-alpha-linked acidic dipeptidase I|FOLH1_HUMAN|Glutamate carboxypeptidase 2|Prostate specific membrane antigen|FGCP|NAALADase I|Pteroylpoly gamma glutamate carboxypeptidase|GIG27|GCP 2|Folh1
- mGCP|Folylpoly gamma glutamate carboxypeptidase|Prostate-specific membrane antigen|Folylpoly-gamma-glutamate carboxypeptidase|Pteroylpoly-gamma-glutamate carboxypeptidase|Membrane glutamate carboxypeptidase|GCP2|Cell growth-inhibiting gene 27 protein|FOLH|GCP II|Glutamate carboxypeptidase II|PSM|Folate hydrolase prostate specific membrane antigen 1|NAALAdase|NAALAD 1|GCPII|NAALAD1|N acetylated alpha linked acidic dipeptidase 1|Glutamate carboxylase II|Folate hydrolase (prostate-specific membrane antigen) 1|Prostate specific membrane antigen variant F|Folate hydrolase|PSMA|Cell growth inhibiting protein 27|FOLH 1|Folate hydrolase 1|N-acetylated-alpha-linked acidic dipeptidase I|FOLH1_HUMAN|Glutamate carboxypeptidase 2|Prostate specific membrane antigen|FGCP|NAALADase I|Pteroylpoly gamma glutamate carboxypeptidase|GIG27|GCP 2|Folh1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q04609
- Q04609
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glutamate carboxypeptidase 2 protein (48-750AA)
- Recombinant Human Glutamate carboxypeptidase 2 protein (48-750AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.