Anti-CELSR2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:10000, IHC:1:25-1:100
Type:
Antigen: CELSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77707-692
- 77707-691
- Antigen Symbol:
- CELSR2
- CELSR2
- Antigen Synonyms:
- CELR2_HUMAN|epidermal growth factor like 2|Multiple epidermal growth factor-like domains 3|Flamingo1|EGFL2|EGF like domain multiple 2|Multiple epidermal growth factor-like domains protein 3|Cadherin family member 10|Celsr2|Cadherin EGF LAG seven-pass G-type receptor 2|MEGF3|CDHF10|Multiple EGF-like domains protein 3|Cadherin EGF LAG seven pass G type receptor 2 flamingo Drosophila homolog|EGF-like protein 2|KIAA0279|Flamingo homolog 3|Epidermal growth factor-like protein 2
- CELR2_HUMAN|epidermal growth factor like 2|Multiple epidermal growth factor-like domains 3|Flamingo1|EGFL2|EGF like domain multiple 2|Multiple epidermal growth factor-like domains protein 3|Cadherin family member 10|Celsr2|Cadherin EGF LAG seven-pass G-type receptor 2|MEGF3|CDHF10|Multiple EGF-like domains protein 3|Cadherin EGF LAG seven pass G type receptor 2 flamingo Drosophila homolog|EGF-like protein 2|KIAA0279|Flamingo homolog 3|Epidermal growth factor-like protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCU4
- Q9HCU4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CELSR2
- Synthetic peptide of Human CELSR2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.