Anti-DIRAS3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:15-1:50
Type:
Antigen: DIRAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77707-643
- 77707-642
- Antigen Symbol:
- DIRAS3
- DIRAS3
- Antigen Synonyms:
- DIRA3_HUMAN|GTP-binding protein Di-Ras3|Rho related GTP binding protein RhoI|DIRAS family GTP binding RAS like 3|NOEY2|GTP-binding RAS-like protein 3|DIRAS3|Distinct subgroup of the Ras family member 3|DIRAS family GTPase 3|Ras homolog gene family member I|RHOI|GTP binding protein Di Ras3|Rho-related GTP-binding protein RhoI|DIRA3|DIRAS family
- DIRA3_HUMAN|GTP-binding protein Di-Ras3|Rho related GTP binding protein RhoI|DIRAS family GTP binding RAS like 3|NOEY2|GTP-binding RAS-like protein 3|DIRAS3|Distinct subgroup of the Ras family member 3|DIRAS family GTPase 3|Ras homolog gene family member I|RHOI|GTP binding protein Di Ras3|Rho-related GTP-binding protein RhoI|DIRA3|DIRAS family
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95661
- O95661
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DIRAS3
- Fusion protein of Human DIRAS3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.