Anti-MCM3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: MCM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77707-516
- 77707-515
- Antigen Symbol:
- MCM3
- MCM3
- Antigen Synonyms:
- P1 Protein|MGC1157|P1.h|p102|MCM3 minichromosome maintenance deficient 3|HCC5|Cervical cancer proto oncogene 5|DNA polymerase alpha holoenzyme-associated protein P1|P1-MCM3|Minichromosome maintenance deficient 3|HCC 5|mcm3|hRlf beta subunit|RLF subunit beta|Replication licensing factor beta subunit|Human cervical cancer proto oncogene 5|RLFB|DNA polymerase alpha holoenzyme associated P1|DNA replication licensing factor mcm3|MCM3_HUMAN|Minichromosome maintenance complex component 3|MCM 3|DNA polymerase alpha holoenzyme associated protein P1|P102 protein|DNA replication factor MCM3|Minichromosome maintenance protein 3|P1 h|P1 MCM3|RLF beta subunit
- P1 Protein|MGC1157|P1.h|p102|MCM3 minichromosome maintenance deficient 3|HCC5|Cervical cancer proto oncogene 5|DNA polymerase alpha holoenzyme-associated protein P1|P1-MCM3|Minichromosome maintenance deficient 3|HCC 5|mcm3|hRlf beta subunit|RLF subunit beta|Replication licensing factor beta subunit|Human cervical cancer proto oncogene 5|RLFB|DNA polymerase alpha holoenzyme associated P1|DNA replication licensing factor mcm3|MCM3_HUMAN|Minichromosome maintenance complex component 3|MCM 3|DNA polymerase alpha holoenzyme associated protein P1|P102 protein|DNA replication factor MCM3|Minichromosome maintenance protein 3|P1 h|P1 MCM3|RLF beta subunit
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25205
- P25205
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MCM3
- Fusion protein of Human MCM3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice.
- Shipped on blue ice.