Anti-SLC39A7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: SLC39A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-347
- 77690-194
- Antigen Symbol:
- SLC39A7
- SLC39A7
- Antigen Synonyms:
- HKE4|Really interesting new gene 5 protein|D6S115E|Zinc transporter SLC39A7|D6S2244E|RING5|HLA class II region expressed gene KE4|H2 KE4|Ke4 gene mouse, human homolog of|SLC39A7|Solute carrier family 39 (zinc transporter), member 7|Histidine-rich membrane protein Ke4|S39A7_HUMAN|KE4|Histidine rich membrane protein Ke4|ZIP7|Solute carrier family 39 member 7|solute carrier family 39 zinc transporter member 7
- HKE4|Really interesting new gene 5 protein|D6S115E|Zinc transporter SLC39A7|D6S2244E|RING5|HLA class II region expressed gene KE4|H2 KE4|Ke4 gene mouse, human homolog of|SLC39A7|Solute carrier family 39 (zinc transporter), member 7|Histidine-rich membrane protein Ke4|S39A7_HUMAN|KE4|Histidine rich membrane protein Ke4|ZIP7|Solute carrier family 39 member 7|solute carrier family 39 zinc transporter member 7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92504
- Q92504
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc transporter SLC39A7 protein (235-380AA)
- Recombinant Human Zinc transporter SLC39A7 protein (235-380AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice