Anti-FOXL1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, WB:1:500-1:2000
Type:
Antigen: FOXL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77720-095
- 77720-094
- Antigen Symbol:
- FOXL1
- FOXL1
- Antigen Synonyms:
- FKH 6|FOX L1|Forkhead box protein L1|FOXL 1|Forkhead-related transcription factor 7|Forkhead box L1|FOXL1_HUMAN|Forkhead related protein FKHL11|Foxl1|FREAC7|Forkhead related transcription factor 7|FREAC-7|Forkhead (Drosophila) like 11|FKHL11|Forkhead-related protein FKHL11|FKHL 11|FREAC 7|FKH6
- FKH 6|FOX L1|Forkhead box protein L1|FOXL 1|Forkhead-related transcription factor 7|Forkhead box L1|FOXL1_HUMAN|Forkhead related protein FKHL11|Foxl1|FREAC7|Forkhead related transcription factor 7|FREAC-7|Forkhead (Drosophila) like 11|FKHL11|Forkhead-related protein FKHL11|FKHL 11|FREAC 7|FKH6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12952
- Q12952
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human FOXL1
- Synthetic peptide of Human FOXL1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice