Anti-KBTBD7 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: KBTBD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77690-191
- 77719-924
- Antigen Symbol:
- KBTBD7
- KBTBD7
- Antigen Synonyms:
- AU045792|Kelch repeat and BTB domain-containing protein 7|KBTBD 7|Kelch repeat and BTB domain containing protein 7|OTTHUMP00000018313|Kelch repeat and BTB (POZ) domain containing 7|DKFZP434E2318|KBTBD7|Gm295|KBTB7_HUMAN|1110008P08Rik
- AU045792|Kelch repeat and BTB domain-containing protein 7|KBTBD 7|Kelch repeat and BTB domain containing protein 7|OTTHUMP00000018313|Kelch repeat and BTB (POZ) domain containing 7|DKFZP434E2318|KBTBD7|Gm295|KBTB7_HUMAN|1110008P08Rik
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WVZ9
- Q8WVZ9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Kelch repeat and BTB domain-containing protein 7 protein (475-684AA)
- Recombinant Human Kelch repeat and BTB domain-containing protein 7 protein (475-684AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice