Anti-ATP6V1C2 Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ATP6V1C2
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77719-701
- 77719-700
- Antigen Symbol:
- ATP6V1C2
- ATP6V1C2
- Antigen Synonyms:
- VMA5|VMA 5|Vacuolar H+ ATPase C2|V-ATPase subunit C 2|ATP6C2|Vacuolar ATP synthase subunit C 2|ATP6V1C2|ATPase H+ transporting lysosomal 42kDa V1 subunit C2|V ATPase subunit C 2|V-type proton ATPase subunit C 2|V type proton ATPase subunit C 2|V ATPase C2 subunit|Vacuolar proton pump subunit C 2|VATC2_HUMAN
- VMA5|VMA 5|Vacuolar H+ ATPase C2|V-ATPase subunit C 2|ATP6C2|Vacuolar ATP synthase subunit C 2|ATP6V1C2|ATPase H+ transporting lysosomal 42kDa V1 subunit C2|V ATPase subunit C 2|V-type proton ATPase subunit C 2|V type proton ATPase subunit C 2|V ATPase C2 subunit|Vacuolar proton pump subunit C 2|VATC2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NEY4
- Q8NEY4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human V-type proton ATPase subunit C 2 protein (190-262AA)
- Recombinant Human V-type proton ATPase subunit C 2 protein (190-262AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice