Anti-CELF6 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CELF6
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-207
- 77719-208
- Antigen Symbol:
- CELF6
- CELF6
- Antigen Synonyms:
- CUGBP Elav-like family member 6|RNA-binding protein BRUNOL-6|RNA binding protein|Bruno like 6|RNA binding protein (Drosophila)|Bruno-like protein 6|6330569O16Rik|CELF6_HUMAN|bruno like 6|CELF-6|RNA binding protein BRUNOL 6|CUG-BP- and ETR-3-like factor 6|CUGBP and ETR3 like factor 6|Celf6|BRUNOL6|Bruno like protein 6
- CUGBP Elav-like family member 6|RNA-binding protein BRUNOL-6|RNA binding protein|Bruno like 6|RNA binding protein (Drosophila)|Bruno-like protein 6|6330569O16Rik|CELF6_HUMAN|bruno like 6|CELF-6|RNA binding protein BRUNOL 6|CUG-BP- and ETR-3-like factor 6|CUGBP and ETR3 like factor 6|Celf6|BRUNOL6|Bruno like protein 6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96J87
- Q96J87
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human CUGBP Elav-like family member 6 protein (292-395AA)
- Recombinant Human CUGBP Elav-like family member 6 protein (292-395AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice