Anti-FAHD2A Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FAHD2A
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77719-161
- 77719-162
- Antigen Symbol:
- FAHD2A
- FAHD2A
- Antigen Synonyms:
- FAH2A_HUMAN|Fumarylacetoacetate hydrolase domain-containing protein 2A|FAHD2A|CGI 105|Fumarylacetoacetate hydrolase domain containing 1|Fumarylacetoacetate hydrolase domain containing 2A|Fumarylacetoacetate hydrolase domain containing protein 2A|FAHD 2A|OTTHUMP00000207352|MGC131995
- FAH2A_HUMAN|Fumarylacetoacetate hydrolase domain-containing protein 2A|FAHD2A|CGI 105|Fumarylacetoacetate hydrolase domain containing 1|Fumarylacetoacetate hydrolase domain containing 2A|Fumarylacetoacetate hydrolase domain containing protein 2A|FAHD 2A|OTTHUMP00000207352|MGC131995
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GK7
- Q96GK7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fumarylacetoacetate hydrolase domain-containing protein 2A protein (4-314AA)
- Recombinant Human Fumarylacetoacetate hydrolase domain-containing protein 2A protein (4-314AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice