Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77719-014
- 77719-013
- Antigen Symbol:
- TP53INP1
- TP53INP1
- Antigen Synonyms:
- p53-dependent damage-inducible nuclear protein 1|Teap|TP53 DINP1|Tumor protein p53-inducible nuclear protein 1|p53 dependent damage inducible nuclear protein 1|FLJ22139|DKFZp434M1317|Stress induced protein|TP53INP1B|p53 inducible p53DINP1|SIP|p53 inducible nuclear protein 1|TP53INP1A|TP53DINP1|TP53INP1|Stress-induced protein|p53DINP1|TP53 INP1|T53I1_HUMAN|Tumor protein p53 inducible nuclear protein 1|p53 damage inducible nuclear protein 1
- p53-dependent damage-inducible nuclear protein 1|Teap|TP53 DINP1|Tumor protein p53-inducible nuclear protein 1|p53 dependent damage inducible nuclear protein 1|FLJ22139|DKFZp434M1317|Stress induced protein|TP53INP1B|p53 inducible p53DINP1|SIP|p53 inducible nuclear protein 1|TP53INP1A|TP53DINP1|TP53INP1|Stress-induced protein|p53DINP1|TP53 INP1|T53I1_HUMAN|Tumor protein p53 inducible nuclear protein 1|p53 damage inducible nuclear protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96A56
- Q96A56
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Tumor protein p53-inducible nuclear protein 1 protein (138-205AA)
- Recombinant Human Tumor protein p53-inducible nuclear protein 1 protein (138-205AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice