Anti-KAT2A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200-1:500
Type:
Antigen: KAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-935
- 77718-936
- Antigen Symbol:
- KAT2A
- KAT2A
- Antigen Synonyms:
- MGC102791|GCN5 general control of amino-acid synthesis 5-like 2 (yeast)|KAT2 A|EC 2.3.1.48|general control of amino acid synthesis 5-like 2|General control of amino acid synthesis, homolog-like 2|hsGCN5|PCAF-b|GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2|KAT2A|1110051E14Rik|AW212720|STAF97|General control of amino acid synthesis protein 5-like 2|Gcn5|GCN5L2|Histone acetyltransferase GCN5|KAT2A_HUMAN|HGCN5|Lysine acetyltransferase 2A|K(lysine) acetyltransferase 2A|Histone acetyltransferase KAT2A|GCN 5
- MGC102791|GCN5 general control of amino-acid synthesis 5-like 2 (yeast)|KAT2 A|EC 2.3.1.48|general control of amino acid synthesis 5-like 2|General control of amino acid synthesis, homolog-like 2|hsGCN5|PCAF-b|GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2|KAT2A|1110051E14Rik|AW212720|STAF97|General control of amino acid synthesis protein 5-like 2|Gcn5|GCN5L2|Histone acetyltransferase GCN5|KAT2A_HUMAN|HGCN5|Lysine acetyltransferase 2A|K(lysine) acetyltransferase 2A|Histone acetyltransferase KAT2A|GCN 5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92830
- Q92830
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histone acetyltransferase KAT2A protein (302-537AA)
- Recombinant Human Histone acetyltransferase KAT2A protein (302-537AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice