Anti-SORL1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SORL1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77718-915
- 77718-916
- Antigen Symbol:
- SORL1
- SORL1
- Antigen Synonyms:
- Sortilin related receptor L(DLR class) A repeats containing|LR 11|C11orf32|SorLA 1|SorLA|SORL_HUMAN|LDLR relative with 11 ligand binding repeats|Mosaic protein LR11|Sortilin-related receptor|LDLR relative with 11 ligand-binding repeats|Sortilin related receptor|LRP9|Sorting protein-related receptor containing LDLR class A repeats|SORL 1|Low-density lipoprotein receptor relative with 11 ligand-binding repeats|SorLA-1|gp250|FLJ21930|FLJ39258|Sorting protein related receptor containing LDLR class A repeats|Low density lipoprotein receptor relative with 11 ligand binding repeats|LRP 9|LR11|SORL1
- Sortilin related receptor L(DLR class) A repeats containing|LR 11|C11orf32|SorLA 1|SorLA|SORL_HUMAN|LDLR relative with 11 ligand binding repeats|Mosaic protein LR11|Sortilin-related receptor|LDLR relative with 11 ligand-binding repeats|Sortilin related receptor|LRP9|Sorting protein-related receptor containing LDLR class A repeats|SORL 1|Low-density lipoprotein receptor relative with 11 ligand-binding repeats|SorLA-1|gp250|FLJ21930|FLJ39258|Sorting protein related receptor containing LDLR class A repeats|Low density lipoprotein receptor relative with 11 ligand binding repeats|LRP 9|LR11|SORL1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92673
- Q92673
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sortilin-related receptor protein (525-744AA)
- Recombinant Human Sortilin-related receptor protein (525-744AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice