Anti-ST8SIA4 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ST8SIA4
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-879
- 77718-878
- Antigen Symbol:
- ST8SIA4
- ST8SIA4
- Antigen Synonyms:
- SIAT8-D|ST8SiaIV|CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase|PST|Polysialyltransferase-1|SIAT8D|PST1|Sialyltransferase 8D|Alpha-2,8-sialyltransferase 8D|EC 2.4.99.-|ST8SIA4|Sialyltransferase St8Sia IV
- SIAT8-D|ST8SiaIV|CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase|PST|Polysialyltransferase-1|SIAT8D|PST1|Sialyltransferase 8D|Alpha-2,8-sialyltransferase 8D|EC 2.4.99.-|ST8SIA4|Sialyltransferase St8Sia IV
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92187
- Q92187
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase protein (21-168AA)
- Recombinant Human CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase protein (21-168AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice