Anti-NFKB2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NFKB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77718-864
- 77718-863
- Antigen Symbol:
- NFKB2
- NFKB2
- Antigen Synonyms:
- Oncogene Lyt 10|H2TF1|Oncogene Lyt-10|Lyt 10|NF kB2|Lymphocyte translocation chromosome 10 protein|Lyt10|Nuclear factor of kappa light polypeptide gene enhancer in B cells 2|DNA binding factor KBF2|Lymphocyte translocation chromosome 10|DNA-binding factor KBF2|p49/p100|Nuclear factor NF kappa B p100 subunit|Nuclear factor of kappa light chain gene enhancer in B cells 2|CVID10|Nuclear factor NF kappa B p52 subunit|p105|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2|NFKB2|Nuclear factor NF-kappa-B p52 subunit|NFKB2_HUMAN
- Oncogene Lyt 10|H2TF1|Oncogene Lyt-10|Lyt 10|NF kB2|Lymphocyte translocation chromosome 10 protein|Lyt10|Nuclear factor of kappa light polypeptide gene enhancer in B cells 2|DNA binding factor KBF2|Lymphocyte translocation chromosome 10|DNA-binding factor KBF2|p49/p100|Nuclear factor NF kappa B p100 subunit|Nuclear factor of kappa light chain gene enhancer in B cells 2|CVID10|Nuclear factor NF kappa B p52 subunit|p105|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2|NFKB2|Nuclear factor NF-kappa-B p52 subunit|NFKB2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00653
- Q00653
- Isotype:
- IgG
- IgG
- Immunogen:
- Synthetic peptide of Human NFKB2
- Synthetic peptide of Human NFKB2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice