Anti-FGFR1 (phospho-Tyr766) Rabbit Polyclonal Antibody
77709-498
New Product
- Antigen Symbol:FGFR1 (phospho-Tyr766)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P11362
- Antigen Synonyms:BFGFR|Fms-like gene|FGFR 1|hydroxyaryl-protein kinase|Proto-oncogene c-Fgr|heparin-binding growth factor receptor|HBGFR|FGFR1/PLAG1 fusion|KAL2|fms-related tyrosine kinase 2|FLT2|FGFR1_HUMAN|Basic fibroblast growth factor receptor 1|FLG|CEK|Fms-like tyrosine kinase 2|HRTFDS|OGD|fibroblast growth factor receptor 1|FLT-2|CD331|FGFR1|HH2|FGFR-1|bFGF-R-1|FGFBR|N-SAM
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of tyrosine 766 (Q-E-Y(p)-L-D) derived from FGFR1
- Purification:Antigen affinity purified
- Cat. No.:77709-498
- Packaging:Vial
WB:1:500-1:1000
Type:
Antigen: FGFR1 (phospho-Tyr766)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat