Anti-CREB1 (phospho-Ser129) Rabbit Polyclonal Antibody
Catalog # 77708-440
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CREB1 (phospho-Ser129)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P16220
- Antigen Synonyms:cAMP response element binding protein 1|MGC9284|cAMP-responsive element-binding protein 1|cAMP responsive element binding protein 1|CREB-1|OTTHUMP00000163864|Transactivator protein|CREB|OTTHUMP00000206660|CREB1|OTTHUMP00000163865|cAMP response element binding protein|CREB1_HUMAN|Cyclic AMP-responsive element-binding protein 1|Active transcription factor CREB|OTTHUMP00000206667|OTTHUMP00000206662
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of serine 129 (I-L-S(p)-R-R) derived from Human CREB
- Purification:Antigen affinity purified
- Cat. No.:77708-440
- Packaging:Vial
Specifications
About this item
WB:1:500-1:1000, IHC:1:50-1:100
Type:
Antigen: CREB1 (phospho-Ser129)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat