Anti-STAC2 Rabbit Polyclonal Antibody
77707-284
New Product
- Antigen Symbol:STAC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6ZMT1
- Antigen Synonyms:STAC2|Src homology 3 and cysteine-rich domain-containing protein 2|24b2/STAC2|SH3 and cysteine-rich domain-containing protein 2
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human STAC2
- Purification:Antigen affinity purified
- Cat. No.:77707-284
- Packaging:Vial
WB:1:500-1:3000
Type:
Antigen: STAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human