Anti-CEP57 Rabbit Polyclonal Antibody
Catalog # 77706-371
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CEP57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q86XR8
- Antigen Synonyms:TSP57|Proliferation inducing protein 8|Cep57 protein|MVA2|FGF2 interacting protein|Centrosomal protein of 57 kDa|Centrosomal protein 57kDa|PIG8|Translokin|KIAA0092|Testis specific protein 57|Cep57|FGF2-interacting protein|CEP57_HUMAN|Testis-specific protein 57
- Storage Buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human CEP57
- Purification:Antigen affinity purified
- Cat. No.:77706-371
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: CEP57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human