Anti-HSPA8 Rabbit Polyclonal Antibody
Catalog # 77705-207
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:HSPA8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P11142
- Antigen Synonyms:HEL 33|HSP73|LPS associated protein|Epididymis luminal protein 33|Heat shock protein 8|Heat shock protein family A (Hsp70) member 8|LPS associated protein 1|HSC71|Heat shock 70kDa protein 8|Heat shock cognate protein 71 kDa|MGC118485|MGC114311|NIP71|HSPA10|Heat shock cognate protein 54|MGC102007|N-myristoyltransferase inhibitor protein 71|MGC131511|Epididymis secretory sperm binding protein Li 72p|Heat shock 70kD protein 8|HSC54|MGC29929|HSP7C_HUMAN|Heat shock 70 kDa protein 8|2410008N15Rik|MGC106514|HSP71|HSPA8|Heat shock 70kD protein 10|Heat-shock70-KD protein 10, formerly|Heat shock protein A8|LAP1|HEL S 72p|Lipopolysaccharide associated protein 1|Hsc73|Heat shock cognate 71 kDa protein|Constitutive heat shock protein 70
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant protein
- Purification:Affinity purified
- Cat. No.:77705-207
- Packaging:Vial
Specifications
About this item
WB:1:1000-2000, IHC:1:200-500
Type:
Antigen: HSPA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat