Anti-ZBTB45 Rabbit Polyclonal Antibody
Catalog # 77705-199
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZBTB45
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q96K62
- Antigen Synonyms:ZNF499|ZBTB45|Zinc finger protein 499|Zinc finger and BTB domain-containing protein 45
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant protein
- Purification:Affinity purified
- Cat. No.:77705-199
- Packaging:Vial
Specifications
About this item
WB:1:500-1000, IHC:1:200-500
Type:
Antigen: ZBTB45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat