Anti-MRPL33 Rabbit Polyclonal Antibody
77705-080
New Product
- Antigen Symbol:MRPL33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:O75394
- Antigen Synonyms:mRpL33|RM33_HUMAN|mitochondrial|MRP-L33|L33mt|39S ribosomal protein L33
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human MRP-L33
- Purification:Antigen affinity purified
- Cat. No.:77705-080
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: MRPL33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human