Anti-ARHGEF2 (phospho-S885) Rabbit Polyclonal Antibody
77705-004
New Product
- Antigen Symbol:ARHGEF2 (phospho-S885)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q92974
- Antigen Synonyms:ARHGEF 2|mKIAA0651|Rho guanine nucleotide exchange factor 2|rho/rac guanine nucleotide exchange factor 2|AA408978|LFP40|Lbcl1|GEF-H1|Protein GEF-H1|ARHGEF2|GEF|KIAA0651|ARHG2_HUMAN|rho/rac guanine nucleotide exchange factor (GEF) 2|Guanine nucleotide exchange factor H1|Microtubule-regulated Rho-GEF|ARHG2|Lfc|Proliferating cell nucleolar antigen p40|GEF H1|MGC95068|rho/rac guanine nucleotide exchange factor|GEFH1|ARHGEF-2|P40
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Lfc around the phosphorylation site of S885
- Purification:Antigen affinity purified
- Cat. No.:77705-004
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: ARHGEF2 (phospho-S885)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat