Anti-NMNAT3 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NMNAT3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-917
- 77721-918
- Antigen Symbol:
- NMNAT3
- NMNAT3
- Antigen Synonyms:
- Pyridine nucleotide adenylyltransferase 3|NMNA3_HUMAN|Nicotinamide mononucleotide adenylyltransferase 3|NMN adenylyltransferase 3|PNAT-3|NMNAT 3|Nmnat3|PNAT3|Nicotinate-nucleotide adenylyltransferase 3|Nicotinamide nucleotide adenylyltransferase 3|PNAT 3|NaMN adenylyltransferase 3
- Pyridine nucleotide adenylyltransferase 3|NMNA3_HUMAN|Nicotinamide mononucleotide adenylyltransferase 3|NMN adenylyltransferase 3|PNAT-3|NMNAT 3|Nmnat3|PNAT3|Nicotinate-nucleotide adenylyltransferase 3|Nicotinamide nucleotide adenylyltransferase 3|PNAT 3|NaMN adenylyltransferase 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96T66
- Q96T66
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3 protein (1-215AA)
- Recombinant Human Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3 protein (1-215AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice