Anti-RUFY1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: RUFY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77721-915
- 77721-914
- Antigen Symbol:
- RUFY1
- RUFY1
- Antigen Synonyms:
- RUFY1_HUMAN|Rab4 interacting protein|Zinc finger FYVE domain-containing protein 12|La-binding protein 1|RABIP4|OTTHUMP00000161521|La binding protein 1|FLJ22251|RUN and FYVE domains containing protein 1|FYVE finger protein EIP1|RUFY1|FYVE-finger protein EIP1|3000002E04Rik|RUN and FYVE domain containing 1|RUN and FYVE domain-containing protein 1|Zinc finger FYVE domain containing protein 12|ZFYVE12|Rab4-interacting protein
- RUFY1_HUMAN|Rab4 interacting protein|Zinc finger FYVE domain-containing protein 12|La-binding protein 1|RABIP4|OTTHUMP00000161521|La binding protein 1|FLJ22251|RUN and FYVE domains containing protein 1|FYVE finger protein EIP1|RUFY1|FYVE-finger protein EIP1|3000002E04Rik|RUN and FYVE domain containing 1|RUN and FYVE domain-containing protein 1|Zinc finger FYVE domain containing protein 12|ZFYVE12|Rab4-interacting protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96T51
- Q96T51
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human RUN and FYVE domain-containing protein 1 protein (439-708AA)
- Recombinant Human RUN and FYVE domain-containing protein 1 protein (439-708AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice