Anti-APIP Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: APIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77721-721
- 77721-720
- Antigen Symbol:
- APIP
- APIP
- Antigen Synonyms:
- APAF1-interacting protein|MMRP19|Apaf1 Interacting Protein|Monocyte/macrophage protein 19|MTRu-1-P dehydratase|CGI 29|CGI29|CG129|MTRu 1 P dehydratase|Probable methylthioribulose-1-phosphate dehydratase|MTNB_HUMAN|dJ179L10.2|APIP|APIP2|Probable methylthioribulose 1 phosphate dehydratase
- APAF1-interacting protein|MMRP19|Apaf1 Interacting Protein|Monocyte/macrophage protein 19|MTRu-1-P dehydratase|CGI 29|CGI29|CG129|MTRu 1 P dehydratase|Probable methylthioribulose-1-phosphate dehydratase|MTNB_HUMAN|dJ179L10.2|APIP|APIP2|Probable methylthioribulose 1 phosphate dehydratase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GX9
- Q96GX9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Methylthioribulose-1-phosphate dehydratase protein (1-242AA)
- Recombinant Human Methylthioribulose-1-phosphate dehydratase protein (1-242AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice