Anti-SGK3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: SGK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-621
- 77721-620
- Antigen Symbol:
- SGK3
- SGK3
- Antigen Synonyms:
- Serum/glucocorticoid regulated kinase 3|Serum/glucocorticoid-regulated kinase-like|SGK 3|Cytokine independent survival kinase|SGK 2|DKFZp781N0293|Serum/glucocorticoid regulated kinase family member 3|SGK3_HUMAN|Sgk3|Serine/threonine-protein kinase Sgk3|SGK2|Serum/glucocorticoid regulated kinase like|CISK|SGKL|Serum/glucocorticoid-regulated kinase 3|Serine/threonine protein kinase Sgk3
- Serum/glucocorticoid regulated kinase 3|Serum/glucocorticoid-regulated kinase-like|SGK 3|Cytokine independent survival kinase|SGK 2|DKFZp781N0293|Serum/glucocorticoid regulated kinase family member 3|SGK3_HUMAN|Sgk3|Serine/threonine-protein kinase Sgk3|SGK2|Serum/glucocorticoid regulated kinase like|CISK|SGKL|Serum/glucocorticoid-regulated kinase 3|Serine/threonine protein kinase Sgk3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96BR1
- Q96BR1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase Sgk3 protein (1-150AA)
- Recombinant Human Serine/threonine-protein kinase Sgk3 protein (1-150AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice