Anti-UPF1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: UPF1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77721-514
- 77721-515
- Antigen Symbol:
- UPF1
- UPF1
- Antigen Synonyms:
- KIAA0221|UPF1 regulator of nonsense transcripts homolog|ATP-dependent helicase RENT1|Smg 2 homolog nonsense mediated mRNA decay factor|Regulator of nonsense transcripts 1|Nonsense mRNA reducing factor 1|NORF1|UPF1 RNA helicase and ATPase|Smg 2|Up-frameshift suppressor 1 homolog|ATP dependent helicase RENT1|FLJ46894|Delta helicase|pNORF1|pNORF 1|RENT1_HUMAN|RENT 1|FLJ43809|NORF 1|Yeast Upf1p homolog|RENT1|UPF 1 regulator of nonsense transcripts homolog|Up frameshift mutation 1 homolog (S. cerevisiae)|HUPF 1|UPF 1|Up frameshift suppressor 1 homolog|hUpf1|Up frameshift mutation 1 homolog|UP Frameshift 1|upf1
- KIAA0221|UPF1 regulator of nonsense transcripts homolog|ATP-dependent helicase RENT1|Smg 2 homolog nonsense mediated mRNA decay factor|Regulator of nonsense transcripts 1|Nonsense mRNA reducing factor 1|NORF1|UPF1 RNA helicase and ATPase|Smg 2|Up-frameshift suppressor 1 homolog|ATP dependent helicase RENT1|FLJ46894|Delta helicase|pNORF1|pNORF 1|RENT1_HUMAN|RENT 1|FLJ43809|NORF 1|Yeast Upf1p homolog|RENT1|UPF 1 regulator of nonsense transcripts homolog|Up frameshift mutation 1 homolog (S. cerevisiae)|HUPF 1|UPF 1|Up frameshift suppressor 1 homolog|hUpf1|Up frameshift mutation 1 homolog|UP Frameshift 1|upf1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92900
- Q92900
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Regulator of nonsense transcripts 1 protein (956-1125AA)
- Recombinant Human Regulator of nonsense transcripts 1 protein (956-1125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice