Anti-OXER1 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OXER1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-243
- 77721-242
- Antigen Symbol:
- OXER1
- OXER1
- Antigen Synonyms:
- TG1019|5 oxo ETE G protein coupled receptor|Oxoeicosanoid receptor 1|Oxoeicosanoid (OXE) receptor 1|G protein coupled receptor R527|G protein coupled receptor TG1019|G-protein coupled receptor 170|5-oxo-ETE G-protein coupled receptor|OTTHUMP00000158809|G-protein coupled receptor TG1019|GPCR|GPR170|G protein coupled receptor 170|G-protein coupled receptor R527|OXER1_HUMAN|5 oxo ETE acid G protein coupled receptor 1|OXER1
- TG1019|5 oxo ETE G protein coupled receptor|Oxoeicosanoid receptor 1|Oxoeicosanoid (OXE) receptor 1|G protein coupled receptor R527|G protein coupled receptor TG1019|G-protein coupled receptor 170|5-oxo-ETE G-protein coupled receptor|OTTHUMP00000158809|G-protein coupled receptor TG1019|GPCR|GPR170|G protein coupled receptor 170|G-protein coupled receptor R527|OXER1_HUMAN|5 oxo ETE acid G protein coupled receptor 1|OXER1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDS5
- Q8TDS5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Oxoeicosanoid receptor 1 protein (1-97AA)
- Recombinant Human Oxoeicosanoid receptor 1 protein (1-97AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice