Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CCDC24 Rabbit Polyclonal Antibody (HRP)
Anti-CCDC24 Rabbit Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CCDC24 Rabbit Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77720-972
  • 77720-973
  • Antigen Symbol:
  • CCDC24
  • CCDC24
  • Antigen Synonyms:
  • CCDC24|Coiled-coil domain-containing protein 24|CCD24_HUMAN|MGC45441|RP5-1198O20.2
  • CCDC24|Coiled-coil domain-containing protein 24|CCD24_HUMAN|MGC45441|RP5-1198O20.2
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8N4L8
  • Q8N4L8
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Coiled-coil domain-containing protein 24 protein (1-307AA)
  • Recombinant Human Coiled-coil domain-containing protein 24 protein (1-307AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications