Anti-PLD6 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: PLD6
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77720-927
- 77720-928
- Antigen Symbol:
- PLD6
- PLD6
- Antigen Synonyms:
- PLD 6|Protein zucchini homolog|Phospholipase D family member 6|Phosphatidylcholine hydrolyzing phospholipase D6|Mitochondrial cardiolipin hydrolase|MitoPLD|Choline phosphatase 6|pld6|Zucchini, Drosophila, homolog of|Phosphatidylcholine-hydrolyzing phospholipase D6|ZUC|Phospholipase D6|Mitochondrial phospholipase|PLD6_HUMAN
- PLD 6|Protein zucchini homolog|Phospholipase D family member 6|Phosphatidylcholine hydrolyzing phospholipase D6|Mitochondrial cardiolipin hydrolase|MitoPLD|Choline phosphatase 6|pld6|Zucchini, Drosophila, homolog of|Phosphatidylcholine-hydrolyzing phospholipase D6|ZUC|Phospholipase D6|Mitochondrial phospholipase|PLD6_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N2A8
- Q8N2A8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitochondrial cardiolipin hydrolase protein (1-252AA)
- Recombinant Human Mitochondrial cardiolipin hydrolase protein (1-252AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice