Anti-RNF125 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RNF125
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-586
- 77720-587
- Antigen Symbol:
- RNF125
- RNF125
- Antigen Synonyms:
- T-cell RING activation protein 1|TRAC-1|EC 2.3.2.27|RNF125|E3 ubiquitin-protein ligase RNF125|RING finger protein 125
- T-cell RING activation protein 1|TRAC-1|EC 2.3.2.27|RNF125|E3 ubiquitin-protein ligase RNF125|RING finger protein 125
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96EQ8
- Q96EQ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase RNF125 protein (143-231AA)
- Recombinant Human E3 ubiquitin-protein ligase RNF125 protein (143-231AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice