Anti-CSH1 Rabbit Polyclonal Antibody
Catalog # 77705-434
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CSH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P01243
- Antigen Synonyms:CSH1|CSH2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthetic peptide from Human protein at AA range: 161-210
- Purification:Antigen affinity purified
- Cat. No.:77705-434
- Packaging:Vial
Specifications
About this item
WB:1:500-10000, ELISA:1:10000
Type:
Antigen: CSH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human