Anti-ROM1 Rabbit Polyclonal Antibody
Catalog # 77689-390
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ROM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q03395
- Antigen Synonyms:ROM|Tspan 23|Tetraspanin-23|Retinal outer segment membrane protein 1|Tetraspanin 23|ROM1_HUMAN|Tspan-23|Rod outer segment membrane protein 1|ROSP1|ROM1|RP7|TSPAN23
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Rod outer segment membrane protein 1 protein (126-263AA)
- Purification:Antigen affinity purified
- Cat. No.:77689-390
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: ROM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse