Anti-ZIP1 Rabbit Polyclonal Antibody
Catalog # 77689-379
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P31111
- Antigen Synonyms:YDR285W|ZIP1|D9819.9Synaptonemal complex protein ZIP1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Saccharomyces cerevisiae Synaptonemal complex protein ZIP1 protein (1-227AA)
- Purification:>95%, Protein G purified
- Cat. No.:77689-379
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: ZIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae