Anti-APOA5 Rabbit Polyclonal Antibody
Catalog # 77713-217
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APOA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6Q788
- Antigen Synonyms:Apoa5|Regeneration associated protein 3|Apolipoprotein A5|RAP3|APOA5_HUMAN|ApoA-V|Regeneration-associated protein 3|Apolipoprotein A-V|Apo-AV|ApoAV
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human APOA5
- Purification:Antigen affinity purified
- Cat. No.:77713-217
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: APOA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human