Anti-FOXE1 Rabbit Polyclonal Antibody
77712-142
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:FOXE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O00358
- Antigen Synonyms:FOX E2|FKHL15|Forkhead box protein E2|FOXE1_HUMAN|FKHL 15|Forkhead (Drosophila) like 15|HFKH4|Forkhead-related protein FKHL15|FOXE1|FOXE2|FKH L15|HFKH 4|Forkhead related protein FKHL 15|HFKL5|Forkhead drosophila homolog like 15|Forkhead box E1|TTF 2|Thyroid transcription factor 2|FOX E1|Forkhead box protein E1|TITF2|FOXE 2|FOXE 1|TTF-2|HFKL 5|Forkhead box E2|TITF 2|Forkhead related protein FKHL15|Forkhead like 15|HNF-3/fork head-like protein 5|TTF2
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human TTF2
- Purification:Antigen affinity purified
- Cat. No.:77712-142
- Packaging:Vial
IF:1:100-1:500
Type:
Antigen: FOXE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse