Anti-MAPK14 (phospho-Thr180) Rabbit Polyclonal Antibody
Catalog # 77711-827
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAPK14 (phospho-Thr180)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q16539
- Antigen Synonyms:p38 MAP kinase|MAX-interacting protein 2|MAP kinase MXI2|p38ALPHA|MAPK14|p38 mitogen activated protein kinase|CSAID-binding protein|p38 MAPK|MAPK 14|Mxi 2|CSAID Binding Protein 1|MAX interacting protein 2|CSPB1|PRKM15|CSBP|CSBP 1|MAP kinase p38 alpha|CSBP2|EXIP|p38 ALPHA|p38alpha Exip|CSBP 2|Mitogen-activated protein kinase 14|p38|Mitogen Activated Protein Kinase 14|RK|MK14_HUMAN|Stress-activated protein kinase 2a|MXI2|MAP kinase 14|Mitogen activated protein kinase p38 alpha|CSBP1|PRKM14|SAPK2A|Csaids binding protein|Mitogen-activated protein kinase p38 alpha|CSAID binding protein|Cytokine suppressive anti-inflammatory drug-binding protein
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of threonine 180 (E-M-T(p)-G-Y ) derived from Human P38MAPK
- Purification:Antigen affinity purified
- Cat. No.:77711-827
- Packaging:Vial
Specifications
About this item
WB:1:500-1:1000, IHC:1:50-1:100
Type:
Antigen: MAPK14 (phospho-Thr180)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat