Anti-TP63 (phospho-Ser395) Rabbit Polyclonal Antibody
Catalog # 77711-565
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:TP63 (phospho-Ser395)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9H3D4
- Antigen Synonyms:OTTHUMP00000209740|p51|Tumor protein p73-like|OTTHUMP00000209734|p73L|Tumor protein p53-like|DN p63 alpha 1|Transformation related protein 63|AIS|OTTHUMP00000209732|Keratinocyte transcription factor KET|OTTHUMP00000209733|OTTHUMP00000209735|Tumor protein p73|P51/P63|OTTHUMP00000209741|OTTHUMP00000209739|Trp63|OTTHUMP00000209743|OFC8|LMS|id:ibd3516|SHFM4|TP63|B(p51A)|p53-related protein p63|p63|Tumor protein 63|EEC3|MGC192897|Keratinocyte transcription factor|KET|OTTHUMP00000209744|p53CP|MGC115972|CUSP|Tumor protein p63 deltaN isoform delta|Trp53rp1|p40|Tumor protein p53-competing protein|OTTHUMP00000209742|Chronic ulcerative stomatitis protein|OTTHUMP00000209737|p73H|Tumor protein p63|Transformation-related protein 63|B(p51B)|TP73L|NBP|TP53CP|TP53L|RHS|OTTHUMP00000209738|P63_HUMAN|Amplified in squamous cell carcinoma|TAp63alpha|DNp63
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of serine 395 (R-R-S(p)-P-D) derived from Human p63
- Purification:Antigen affinity purified
- Cat. No.:77711-565
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TP63 (phospho-Ser395)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat